Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(III) acetate hydrate

Catalog No. 367753250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-775-3250G 250 g
36-775-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-775-3250G Supplier Sigma Aldrich Supplier No. 367753250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.Cerium (III) acetate hydrate may be used as the starting material for the synthesis of CeO2 nanoparticles.

Specifications

CAS 206996-60-3
Quantity 250 g
Molecular Formula C6H9CeO6 -+ xH2O
Linear Formula Ce(CH3CO2)3 -+ xH2O
MDL Number MFCD00150533
Molecular Weight (g/mol) 317.25
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.