Learn More
Strem, An Ascensus Company CAS# 54451-25-1. 50g. Cerium(III) carbonate hydrate (99+%-Ce) (REO). MFCD00149635
SDP
Catalog No. 5090115877
Encompass
CAS# 54451-25-1. 50g. Cerium(III) carbonate hydrate (99+%-Ce) (REO). MFCD00149635. Molecular Weight: 460.29. Molecular Formula: Ce2(CO3)3∙XH2O. Color/form: white xtl. Strem# 93-5807. http://www.strem.com/catalog/v/93-5807/
Catalog No. 50-901-15877 Supplier Strem, An Ascensus Company Supplier No. 93580750G
CAS# 54451-25-1. 50g. Cerium(III) carbonate hydrate (99+%-Ce) (REO). MFCD00149635. Molecular Weight: 460.29. Molecular Formula: Ce2(CO3)3∙XH2O. Color/form: white xtl. Strem# 93-5807. http://www.strem.com/catalog/v/93-5807/