Learn More
Strem, An Ascensus Company CAS# 7790-86-5. 5g. Cerium(III) chloride, anhydrous (H2O < 0.5%) (99.9%-Ce) (REO). MFCD00010929
SDP
Catalog No. 501385565
Encompass
CAS# 7790-86-5. 5g. Cerium(III) chloride, anhydrous (H2O < 0.5%) (99.9%-Ce) (REO). MFCD00010929. Molecular Weight: 246.48. Molecular Formula: CeCl3. Color/form: white pwdr. Strem# 93-5862. http://www.strem.com/catalog/v/93-5862/
Catalog No. 50-138-5565 Supplier Strem, An Ascensus Company Supplier No. 9358625G
CAS# 7790-86-5. 5g. Cerium(III) chloride, anhydrous (H2O < 0.5%) (99.9%-Ce) (REO). MFCD00010929. Molecular Weight: 246.48. Molecular Formula: CeCl3. Color/form: white pwdr. Strem# 93-5862. http://www.strem.com/catalog/v/93-5862/