Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(III) Chloride Heptahydrate

Catalog No. 22300100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-300-100G 100 g
22-300-500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-300-100G Supplier Sigma Aldrich Supplier No. 22300100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cerium(III) chloride heptahydrate is a mild Lewis acid. It participates in the allylation of aldehydes. Cerium(III) chloride heptahydrate in combination with sodium iodide supported on silica gel efficiently promotes Michael-type addition. Cerium(III) chloride heptahydrate in the presence of sodium iodide acts as an efficient catalyst for the Michael addition of 1,3-dicarbonyl compounds to +a,+a-unsaturated ketones and +a,+a-unsaturated aldehydes.Cerium(III) chloride heptahydrate was used for the preparation of undoped ceria nanoparticles. It may be used in the preparation of 2-indolyl-1-nitroalkane derivatives and CeO2-supported Co catalysts.

Specifications

CAS 18618-55-8
Density ∼3.94 g/mL at 25°C
Molecular Formula CeCl3 -+ 7H2O
Linear Formula CeCl3 -+ 7H2O
MDL Number MFCD00149634
Quantity 100 g
Synonym Cerium trichloride heptahydrate; Cerous chloride heptahydrate
Molecular Weight (g/mol) 372.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.