Learn More
Strem, An Ascensus Company CAS# 19423-76-8. 100g. Cerium(III) chloride hydrate (99.9%-Ce) (REO). MFCD00149633
SDP
Catalog No. 5090115879
Encompass
CAS# 19423-76-8. 100g. Cerium(III) chloride hydrate (99.9%-Ce) (REO). MFCD00149633. Molecular Weight: 246.48. Molecular Formula: CeCl3∙XH2O. Color/form: white to pale yellow xtl. Strem# 93-5808. http://www.strem.com/catalog/v/93-5808/
Catalog No. 50-901-15879 Supplier Strem, An Ascensus Company Supplier No. 935808100G
CAS# 19423-76-8. 100g. Cerium(III) chloride hydrate (99.9%-Ce) (REO). MFCD00149633. Molecular Weight: 246.48. Molecular Formula: CeCl3∙XH2O. Color/form: white to pale yellow xtl. Strem# 93-5808. http://www.strem.com/catalog/v/93-5808/