Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) chloride, ultra dry, 99.9% (REO)

Catalog No. AA3568822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3568822 100 g
AA3568806 5 g
AA3568814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3568822 Supplier Thermo Scientific Chemicals Supplier No. 03568822
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7790-86-5 | CeCl3 | 246.466 g/mol

Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.466
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Boiling Point 1727°C
Melting Point 848°C
Color White
Physical Form Crystalline
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline beads
Assay Percent Range 99.9%
Density 3.92
Solubility Information Soluble in water,ethanol,acids and acetone.
Formula Weight 246.48
Chemical Name or Material Cerium(III) chloride
Show More Show Less
Hazard Category H314
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-227-8
RTECSNumber FK5075000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.