Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. AA4140818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4140818 50 g
AA4140809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4140818 Supplier Thermo Scientific Chemicals Supplier No. 04140818
Add to Cart

missing translation for 'validMainframeMsg'

For vacuum deposition | CAS: 7758-88-5 | CeF3 | 197.111 g/mol

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-88-5
Molecular Formula CeF3
Molecular Weight (g/mol) 197.111
MDL Number MFCD00010932
InChI Key QCCDYNYSHILRDG-UHFFFAOYSA-K
Synonym cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
PubChem CID 24457
IUPAC Name cerium(3+);trifluoride
SMILES [F-].[F-].[F-].[Ce+3]

Specifications

Boiling Point ∼2300°C
Melting Point 1460°C
Physical Form Pieces
Quantity 50 g
Merck Index 14,1998
Formula Weight 197.12
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.16 g/mL
Assay (REO)
Chemical Name or Material Cerium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 231-841-3
RTECSNumber FK6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.