Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) and Dilute Nitric acid, Etchant Solution, Ce(IV) concentration 0.25N, Thermo Scientific Chemicals

Catalog No. AA42110K7
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42110K7 4 L
AA42110K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42110K7 Supplier Thermo Scientific Chemicals Supplier No. 042110K7
Add to Cart

missing translation for 'validMainframeMsg'

It is widely applied as etchant solution in industrial purpose to chemically remove layers from the surface of a wafer during manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01866052

Specifications

Physical Form Liquid
Quantity 4 L
UN Number UN3098
Solubility Information Soluble in water
Concentration or Composition (by Analyte or Components) Ce(IV) concentration 0.25N
Chemical Name or Material Cerium(IV) and Dilute Nitric acid, Etchant Solution
Hazard Category H290-H314-H317-H335
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.