Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(Iv) Fluoride

Catalog No. 4359375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-593-75G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 43-593-75G Supplier Sigma Aldrich Supplier No. 4359375G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Synthesis of sodium-rare earth silicate oxide-fluoride apatitesC60 fluorination with rare earth metal tetrafluoridesSynthesis of cerium(IV) fluoride compoundsReagent for:Fluorination of fluorocyclobutene with high-valency metal fluorideCatalyst for Hantzsch reactionUsed for stain etching to form microporous silicon

Specifications

CAS 26115-70-8
Density 1.17 g/mL at 25°C
Boiling Point 250°C
Molecular Formula C21H45N3O12Si3
Linear Formula C21H45N3O12Si3
MDL Number MFCD00054746
Quantity 100 mL
Synonym Ceric fluoride; Cerium tetrafluoride
Molecular Weight (g/mol) 615.85 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.