Learn More
Strem, An Ascensus Company CAS# 1306-38-3. 50g. Cerium(IV) oxide (99.9%-Ce) (REO). MFCD00010933
SDP
Catalog No. 5090115887
Encompass
CAS# 1306-38-3. 50g. Cerium(IV) oxide (99.9%-Ce) (REO). MFCD00010933. Molecular Weight: 172.12. Molecular Formula: CeO2. Color/form: yellow pwdr. Strem# 93-5816. http://www.strem.com/catalog/v/93-5816/
Catalog No. 50-901-15887 Supplier Strem, An Ascensus Company Supplier No. 93581650G
CAS# 1306-38-3. 50g. Cerium(IV) oxide (99.9%-Ce) (REO). MFCD00010933. Molecular Weight: 172.12. Molecular Formula: CeO2. Color/form: yellow pwdr. Strem# 93-5816. http://www.strem.com/catalog/v/93-5816/