Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(IV) oxide-gadolinium doped

Catalog No. 57235725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-235-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-235-725G Supplier Sigma Aldrich Supplier No. 57235725G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gadolinium doped ceria may be prepared by cation complexation sol gel technique. XRD, specific surface area, morphology has been described in a study. Higher oxygen ion conductivity of GDC over zirconia based materials makes it a preferable choice for some components of SOFCs.Gadolinium doped ceria (GDC) may be used as an intermediate temperature electrolyte for solid oxide fuel cells (SOFCs) at elevated temperatures.

Specifications

Molecular Formula CeO2
Linear Formula CeO2
Quantity 25 g
MDL Number MFCD04040745
Synonym Gadolinium doped ceria
Molecular Weight (g/mol) 172.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.