Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(IV) oxide-yttria doped

Catalog No. 76375625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-375-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-375-625G Supplier Sigma Aldrich Supplier No. 76375625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cerium(IV) oxide-yttria doped (YDC-20) is an electrolytic material that is used as an alternative to zirconia-based electrolytes. Its properties include high ionic conductivity (0.01 S cm1), electronic conductivity (3 S cm1), enhanced electrode reactions, and high oxygen surface exchange coefficient.YDC-20 can be used as an electrolyte in the fabrication of solid oxide fuel cells (SOFCs). A thin film of YDC-20 (∼17.5 nm) effectively reduces the cathode/interfacial energy by 0.1 eV and increases the exchange current density by a factor of 4. It can be operated with a temperature range of 300°C to 500°C.

Specifications

CAS 170033-03-1
Molecular Formula CeO2/Y2O3
Linear Formula CeO2/Y2O3
Quantity 25 g
MDL Number MFCD04039918
Synonym 20 YDC; YDC-20; YDC20; Cerium yttrium oxide; YDC; Yttria doped ceria; Yttrium doped ceria
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.