Learn More
Abcam Cerivastatin sodium, HMG-CoA reductase inhibitor, 25MG
Catalog No. 502706157
Encompass_Preferred
MW 481.5 Da, Purity >98%. Potent HMG-CoA reductase inhibitor (Ki = 13 nM). Shows higher potency than Lovastatin (ab120614). Enantiomerically pure pyridine derivative. Reduces plasma cholesterol levels in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6157 Supplier Abcam Supplier No. AB14285325MG
Add to Cart
Add to Cart
MW 481.5 Da, Purity >98%. Potent HMG-CoA reductase inhibitor (Ki = 13 nM). Shows higher potency than Lovastatin (ab120614). Enantiomerically pure pyridine derivative. Reduces plasma cholesterol levels in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement