Learn More
Abcam Cerivastatin sodium, HMG-CoA reductase inhibitor, 5MG
Catalog No. 502706156
Encompass_Preferred
MW 481.5 Da, Purity >98%. Potent HMG-CoA reductase inhibitor (Ki = 13 nM). Shows higher potency than Lovastatin (ab120614). Enantiomerically pure pyridine derivative. Reduces plasma cholesterol levels in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6156 Supplier Abcam Supplier No. AB1428535MG
MW 481.5 Da, Purity >98%. Potent HMG-CoA reductase inhibitor (Ki = 13 nM). Shows higher potency than Lovastatin (ab120614). Enantiomerically pure pyridine derivative. Reduces plasma cholesterol levels in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement