Learn More
Aviva Systems Biology Corp CERKL Antibody - C-terminal region (ARP59827_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CERKL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CERKL. Alternate names: RP26ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-847