Learn More
PDC Cerner-Compatible Direct Thermal Paper Labels, 1000 Labels/Roll, 6 Rolls/Box
Catalog No. 21235824
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 6 Rolls/Box
Catalog No. Quantity
21-235-824 1000 Labels/Roll, 6 Rolls/Box
1 options
Catalog No. 21-235-824 Supplier PDC Supplier No. TD1PN8
Add to Cart
Add to Cart
Cerner-compatible direct thermal paper labels with permanent adhesive, ideal for clinical and laboratory use.
  • Compatible with Cerner systems for accurate patient and specimen labeling
  • Direct thermal paper prints clearly without the need for ink or toner
  • Permanent adhesive keeps labels securely in place during use
  • Label format supports clear text and barcode readability
  • 1 in. core works with standard label printers
  • Supplied in bulk with 1000 labels per roll and 6 rolls per box
  • Suitable for everyday use in clinical and laboratory settings
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 6 Rolls/Box
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm