Learn More
PDC Cerner Direct Thermal Paper Label, 1000 Labels/Roll, 2 Rolls/Case
Catalog No. 21235793
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 2 Rolls/Cs.
Catalog No. Quantity
21-235-793 1000 Labels/Roll, 2 Rolls/Cs.
1 options
Catalog No. 21-235-793 Supplier PDC Supplier No. LDWHP117
Add to Cart
Add to Cart
Cerner direct thermal paper labels with permanent adhesive for clear, durable printing and accurate clinical labeling.
  • Permanent adhesive provides dependable attachment for clinical workflows
  • White face stock ensures high contrast for legible, easy-to-read printing of patient and specimen information
  • Optimized label format supports efficient documentation and organization in healthcare settings
  • Compatible with standard label dispensers and printers, enabling seamless use across facilities
  • Engineered for consistent, accurate, and durable labeling in demanding clinical environments
  • Bulk packaging with 1000 labels per roll and 2 rolls per case helps maintain continuous labeling operations
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 2 Rolls/Cs.
Dimensions (L x W) 4 x 4 1/2 in.
Length (English) 4.5 in.
Width (English) 4 in.
Width (Metric) 101.6 mm