Learn More
PDC Cerner Direct Thermal Paper Label Permanent, 3000/Roll, 6 Rolls/Box
Catalog No. 21235841
Change view
Click to view available options
Quantity:
3000/Roll, 6 Rolls/Box
Catalog No. Quantity
21-235-841 3000/Roll, 6 Rolls/Box
1 options
Catalog No. 21-235-841 Supplier PDC Supplier No. TDPN8
  • $276.00
    $204.65 / Case of 6
    Save $71.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
High-volume Cerner-compatible direct thermal labels with permanent adhesive, designed for clear and reliable patient identification.
  • Direct thermal paper for ribbon-free printing
  • Compact 2 in. x 1 1/4 in. size for versatile labeling
  • Compatible with Cerner systems for seamless integration
  • Permanent adhesive ensures long-lasting label placement
  • 3 in. core fits standard thermal printers
  • 3000 labels per roll, 6 rolls per box for large-scale needs
  • White background for high-contrast printing
  • Crisp barcode and text clarity to reduce errors
  • Supports patient safety through accurate specimen labeling
  • Ideal for healthcare and laboratory environments
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 3000/Roll, 6 Rolls/Box
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm