Learn More
PDC Cerner Direct Thermal Paper Label, Yellow, 1000 Labels/Roll, 2 Rolls/Case
Catalog No. 21235792
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 2 Rolls/Cs.
Catalog No. Quantity
21-235-792 1000 Labels/Roll, 2 Rolls/Cs.
1 options
Catalog No. 21-235-792 Supplier PDC Supplier No. LDWHP101
Add to Cart
Add to Cart
Cerner yellow labels with permanent adhesive for clear, durable printing and efficient healthcare labeling.
  • Permanent adhesive ensures dependable attachment for accurate Cerner system labeling
  • Bright yellow color improves visibility and helps quickly identify patient or specimen information
  • Optimized label format supports clear documentation and organization in clinical workflows
  • Compatible with standard industrial dispensers, enabling seamless use in healthcare facilities
  • Engineered for consistent, precise, and durable labeling under demanding healthcare conditions
  • Packaged with 1000 labels per roll and 2 rolls per case to meet high-volume labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color Yellow
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 2 Rolls/Cs.
Dimensions (L x W) 4 x 4 1/2 in.
Length (English) 4 in.
Width (English) 4.5 in.
Width (Metric) 114.3 mm