Learn More
Thermco Products Inc CERT W/THERM 0,100,101C POINTS
Catalog No. NC1141795
Encompass_Preferred
ASTM 121C thermometer (Hg) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC1141795 Supplier Thermco Products Inc Supplier No. ACC121CFC
ASTM 121C thermometer (Hg) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.