Learn More
Certified Safety Certi-Cool Junior Instant Cold Packs
SDP
Catalog No. 19808886
Change view
Click to view available options
Dimensions (L x W):
5 x 7 in.
Catalog No. Dimensions (L x W)
19-808-886 5 x 7 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-808-886 Supplier Certified Safety Supplier No. 216002
This item is not returnable. View return policy
Specifications
Product Type Cold Pack
For Use With (Application) Treatment of swelling, burns, nose bleeds and headaches
Quantity 25 Pk.
Dimensions (L x W) 5 x 7 in.
To Activate Cold Pack; Punch/Pop Inner bag to Activate