Learn More
Waters Corp CERTIFIED 15 X 75 MM CLEAR GLA
Supplier:  Waters Corp 186004903C
Encompass_Preferred
Certified 15 x 75 mm Clear Glass Screw Cap Vial with Cap and Bonded Pre-slit PTFE/Silicone Septum 100 pack for use in Patrol Acquity UPLC,PTFE/silicone Septum,Clear Glass Vial,Glass Type: Type 1 Class A,
Catalog No. 50-791-224