Learn More
Waters Corp WATERS CORPORATION Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 2 mL, Septumless Cap
Supplier:  Waters Corp 186004132C
Encompass_Preferred
Certified vials are tested for cleanliness by HPLC and maintained to the industry’s tightest dimensional tolerances. LCGC Certified Clear Glass Vials undergo UV-testing by HPLC to detect trace levels of chemicals from manufacturing and packaging. The vial features a universal screw neck design, forming an excellent seal, and a septumless cap suitable for single injection and MS applications due to its excellent solvent resistance and chemical compatibility. Made from type 1, 33-expansion glass, the most chemically inert glass available, these 2 mL vials ensure high-quality test results.
  • Tested for cleanliness by HPLC.
  • Maintained to tightest dimensional tolerances.
  • Universal screw neck design for excellent seal.
  • Septumless cap for single injection and MS applications.
  • Made from type 1, 33-expansion glass, the most chemically inert glass.
Catalog No. 50-782-557