Learn More
Waters Corp Autosampler Vial, LCGC Certified, Amber Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50782558 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These LCGC Certified Amber Glass 12 x 32 mm Screw Neck Vials, with Polyethylene Septumless Cap, 2 mL Volume, are sold in a pack of 100. They are tested for cleanliness by HPLC and held to tight dimensional tolerances, offering superior performance and sample integrity for liquid and gas chromatography systems. Crafted from high-quality borosilicate glass, these vials provide robust durability and resistance against breakage and contamination. The polyethylene septumless cap ensures a secure seal, preventing sample loss and simplifying sample loading.
  • LCGC-certified for cleanliness and compliance with liquid and gas chromatography standards.
  • Secure polyethylene septumless cap prevents sample loss.
  • Suitable for liquid chromatography, gas chromatography, sample storage, and analysis.
Catalog No. 50-782-558 Supplier Waters Corp Supplier No. 186004133C
These LCGC Certified Amber Glass 12 x 32 mm Screw Neck Vials, with Polyethylene Septumless Cap, 2 mL Volume, are sold in a pack of 100. They are tested for cleanliness by HPLC and held to tight dimensional tolerances, offering superior performance and sample integrity for liquid and gas chromatography systems. Crafted from high-quality borosilicate glass, these vials provide robust durability and resistance against breakage and contamination. The polyethylene septumless cap ensures a secure seal, preventing sample loss and simplifying sample loading.
  • LCGC-certified for cleanliness and compliance with liquid and gas chromatography standards.
  • Secure polyethylene septumless cap prevents sample loss.
  • Suitable for liquid chromatography, gas chromatography, sample storage, and analysis.