Learn More
Waters Corp Waters Corporation Container, LDPE, 1000 mL
Catalog No. 502038912 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Certified LDPE 1000 mL Containers are specifically designed for use with Waters BioResolve CX Buffer Concentrates. Constructed from low-density polyethylene, this container is soft for easy squeezing and dispensing. Each container undergoes strict quality control testing for cleanliness, ensuring reliable performance in laboratory settings.
  • Designed specifically for use with BioResolve CX Buffer Concentrates.
  • Made of low-density polyethylene for ease of use and compatibility.
  • Soft and can be squeezed without causing damage or stress cracks.
  • Strong resistance to chemicals.
  • Offers a good vapor barrier.
  • Strictly QC tested for cleanliness.
Catalog No. 50-203-8912 Supplier Waters Corp Supplier No. 186009110
Waters Certified LDPE 1000 mL Containers are specifically designed for use with Waters BioResolve CX Buffer Concentrates. Constructed from low-density polyethylene, this container is soft for easy squeezing and dispensing. Each container undergoes strict quality control testing for cleanliness, ensuring reliable performance in laboratory settings.
  • Designed specifically for use with BioResolve CX Buffer Concentrates.
  • Made of low-density polyethylene for ease of use and compatibility.
  • Soft and can be squeezed without causing damage or stress cracks.
  • Strong resistance to chemicals.
  • Offers a good vapor barrier.
  • Strictly QC tested for cleanliness.