Learn More
Fisherbrand™ Certified Low-Particulate Boston Round Bottles
GREENER_CHOICE
Catalog No. 02911889 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-911-889 125 mL
02-911-892 250 mL
02-911-879 500 mL
02-911-914 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-911-889
Add to Cart
Add to Cart
Ideal storage for solvents, chemicals and samples. Fisherbrand™ Certified Low Particulate Boston Round Bottles are manufactured from clear soda glass.
  • Narrow mouth, clear glass
  • Certified to be cleaned for low particulates, analysis certificate attached
  • Five particles or less per mL with WFI in a class 100/10 cleanroom
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
Cleaning Procedure USP purified Water
Color Clear
Material Glass
Quantity 24/PK.
Closure Color White
Liner Material Polyvinyl
Closure Included Yes
Mouth Wide
Closure Liner Polyvinyl
Closure Material PTFE
Closure Size 22-400 mm
Closure Type Screw Cap
Shape Round
Certifications/Compliance Certified low particulate
Product Type Boston Round Bottle