Learn More
Chemglass Life Sciences Certified Pack, 2.0mL Clear with Grad Spot 12x32 9mm Thread Large Opening Vial and 0.040" PTFE/Silicone Septa Lined Blue Bonded Closure
SDP
Catalog No. 5000980706
Encompass_Preferred
Certified Pack, 2.0mL Clear with Grad Spot 12x32 9mm Thread Large Opening Vial and 0.040" PTFE/Silicone Septa Lined Blue Bonded Closure
Catalog No. 50-009-80706 Supplier Chemglass Life Sciences Supplier No. CV5323CERT
Certified Pack, 2.0mL Clear with Grad Spot 12x32 9mm Thread Large Opening Vial and 0.040" PTFE/Silicone Septa Lined Blue Bonded Closure