Learn More
MilliporeSigma™ Supelco™ Certified Polypropylene Bonded Caps with Septa
Catalog No. 111011713 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Septum:
Bonded/Silicone
Rubber
Product Type:
Hole Cap
Hole Cap (bonded septa)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum Product Type
11-101-1713 Bonded/Silicone Hole Cap (bonded septa)
11-101-2014 Rubber Hole Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1713 Supplier MilliporeSigma™ Supelco™ Supplier No. 29320U
Add to Cart
Add to Cart
Bonded caps with septa
  • Manufactured using virgin resins to eliminate the possibility of sample contamination.
  • Caps are molded to ensure consistency in fit, thread specification and overall dimensions.
  • The PTFE/silicone septa are manufactured, conditioned and then tested by GC/MS to ensure they have the lowest level of extractable in the industry.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Cap 9 mm
Color Dark Blue
Material PTFE, Polypropylene
Pre-slit Yes
Septum Bonded/Silicone
No. per Pack 100
Product Type Hole Cap (bonded septa)