Learn More
Waters Corp Waters Vac Cartridge, Alumina A, 6 cc, 500 mg Sorbent
Catalog No. 50791151 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Certified Sep-Pak Alumina A 6 cc Vac Cartridge is specifically engineered for solid phase extraction applications, utilizing 500 mg of Alumina A sorbent. It provides reliable performance for various laboratory separations, designed for efficiency and compatibility with modern analytical techniques.
  • Solid phase extraction product type.
  • Features Alumina A chemistry.
  • Not endcapped.
  • Vac cartridge format.
  • Mass spectrometry compatible.
  • Operates in normal-phase mode.
  • Particle size ranges from 50 to 300 μm.
  • Pore size is 120 Å.
  • Alumina sorbent substrate.
  • Sorbent weight of 500 mg.
  • Water wettable.
  • Supports a pH range of 0 to 10.
  • Barrel size is 6 cc.
  • Hold up volume is 0.5 mL.
  • Available in packs of 30.
Catalog No. 50-791-151 Supplier Waters Corp Supplier No. 186004603
This Certified Sep-Pak Alumina A 6 cc Vac Cartridge is specifically engineered for solid phase extraction applications, utilizing 500 mg of Alumina A sorbent. It provides reliable performance for various laboratory separations, designed for efficiency and compatibility with modern analytical techniques.
  • Solid phase extraction product type.
  • Features Alumina A chemistry.
  • Not endcapped.
  • Vac cartridge format.
  • Mass spectrometry compatible.
  • Operates in normal-phase mode.
  • Particle size ranges from 50 to 300 μm.
  • Pore size is 120 Å.
  • Alumina sorbent substrate.
  • Sorbent weight of 500 mg.
  • Water wettable.
  • Supports a pH range of 0 to 10.
  • Barrel size is 6 cc.
  • Hold up volume is 0.5 mL.
  • Available in packs of 30.