Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Alumina A, 500 mg Sorbent, 50-300 μm
Catalog No. 50791150 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Certified Sep-Pak Alumina A 3 cc Vac Cartridge, designed for solid phase extraction. It contains 500 mg of Alumina A sorbent with a particle size of 50 to 300 μm. It is compatible with mass spectrometry and is water wettable. The cartridge has a barrel size of 3 cc and a hold-up volume of 0.4 mL.
  • Mass spec compatibility: yes
  • Water wettable: yes
  • Mode: normal-phase
  • PH range: 0 to 10 pH
Catalog No. 50-791-150 Supplier Waters Corp Supplier No. 186004602
This product is a Certified Sep-Pak Alumina A 3 cc Vac Cartridge, designed for solid phase extraction. It contains 500 mg of Alumina A sorbent with a particle size of 50 to 300 μm. It is compatible with mass spectrometry and is water wettable. The cartridge has a barrel size of 3 cc and a hold-up volume of 0.4 mL.
  • Mass spec compatibility: yes
  • Water wettable: yes
  • Mode: normal-phase
  • PH range: 0 to 10 pH