Learn More
Waters Corp WATERS CORPORATION Sep-Pak Vac Cartridge, Alumina B, 6 cc, 500 mg
Catalog No. 50791154 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Certified Sep-Pak™ cartridges are disposable solid-phase extraction (SPE) devices for convenient and reproducible sample preparation. Manufactured in an ISO 13485 and ISO 9002-registered facility, they adhere to cGMP guidelines for Class 1 Medical Devices.
  • Supports manual and automated solid-phase extraction.
  • Available in 3 cc and 6 cc formats with Reversed-Phase (C18) and Normal-Phase (Silica, Alumina A, Alumina B, Alumina N, Florisil™) chemistries.
  • Ensures stability when stored in original sealed pouch; utilizes highly active sorbents.
  • Recommended conditioning flow rate: 5 to 10 mL/min.
  • Loading/eluting flow rates vary by cartridge type, from 0.2 to 1 mL/min to 2 to 10 mL/min.
  • Capacity is dependent on sample properties, sorbent type/quality, and retention factor (k).
Catalog No. 50-791-154 Supplier Waters Corp Supplier No. 186004606
Certified Sep-Pak™ cartridges are disposable solid-phase extraction (SPE) devices for convenient and reproducible sample preparation. Manufactured in an ISO 13485 and ISO 9002-registered facility, they adhere to cGMP guidelines for Class 1 Medical Devices.
  • Supports manual and automated solid-phase extraction.
  • Available in 3 cc and 6 cc formats with Reversed-Phase (C18) and Normal-Phase (Silica, Alumina A, Alumina B, Alumina N, Florisil™) chemistries.
  • Ensures stability when stored in original sealed pouch; utilizes highly active sorbents.
  • Recommended conditioning flow rate: 5 to 10 mL/min.
  • Loading/eluting flow rates vary by cartridge type, from 0.2 to 1 mL/min to 2 to 10 mL/min.
  • Capacity is dependent on sample properties, sorbent type/quality, and retention factor (k).