Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Alumina N, 3 cc, 500 mg
Catalog No. 50791156 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Sep-Pak Vac cartridges provide normal-phase solid phase extraction with highly active alumina. They offer enhanced stability in high pH conditions and ensure reproducible separations and superior performance for ultra-low level analyte detection.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention.
  • More stable than unfunctionalized silica under high pH.
  • Quality controlled for reproducible separations.
  • Superior performance for ultra-low level analyte detection.
  • Syringe-barrel design for vacuum manifolds and automated SPE.
  • Manufactured in ISO 13485/ISO 9002-registered facility, complying with cGMP for Class 1 Medical Devices.
  • Suitable for manual and automated SPE.
Catalog No. 50-791-156 Supplier Waters Corp Supplier No. 186004608
These Sep-Pak Vac cartridges provide normal-phase solid phase extraction with highly active alumina. They offer enhanced stability in high pH conditions and ensure reproducible separations and superior performance for ultra-low level analyte detection.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention.
  • More stable than unfunctionalized silica under high pH.
  • Quality controlled for reproducible separations.
  • Superior performance for ultra-low level analyte detection.
  • Syringe-barrel design for vacuum manifolds and automated SPE.
  • Manufactured in ISO 13485/ISO 9002-registered facility, complying with cGMP for Class 1 Medical Devices.
  • Suitable for manual and automated SPE.