Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, tC18, 3 cc, 500 mg
Catalog No. 50791166 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Certified Sep-Pak tC18 3 cc Vac Cartridges are convenient, reproducible, disposable solid phase extraction (SPE) devices for sample preparation. They simplify sample extraction and preparation, facilitating transfer of solvents, solutions, and samples without spillage or loss. Manufactured in a Waters ISO 13485 and ISO 9002-registered facility, compliant with cGMP for Class 1 Medical Devices.
  • Silica-based bonded phase, strong hydrophobicity, and trifunctional bonding chemistry for increased hydrolytic stability.
  • Provides unique selectivity, excellent analyte recovery, and is recommended for both polar and non-polar analytes.
  • Quality controlled for reproducible separations and superior performance in ultra-low level analyte detection.
  • Syringe-barrel-type cartridges for vacuum manifolds and automated SPE instruments.
Catalog No. 50-791-166 Supplier Waters Corp Supplier No. 186004619
Certified Sep-Pak tC18 3 cc Vac Cartridges are convenient, reproducible, disposable solid phase extraction (SPE) devices for sample preparation. They simplify sample extraction and preparation, facilitating transfer of solvents, solutions, and samples without spillage or loss. Manufactured in a Waters ISO 13485 and ISO 9002-registered facility, compliant with cGMP for Class 1 Medical Devices.
  • Silica-based bonded phase, strong hydrophobicity, and trifunctional bonding chemistry for increased hydrolytic stability.
  • Provides unique selectivity, excellent analyte recovery, and is recommended for both polar and non-polar analytes.
  • Quality controlled for reproducible separations and superior performance in ultra-low level analyte detection.
  • Syringe-barrel-type cartridges for vacuum manifolds and automated SPE instruments.