Learn More
Waters Corp Waters Vac Cartridge, Florisil, 3 cc, 500 mg Sorbent, 50-200 μm
Catalog No. 50791159 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These convenient, reproducible, and disposable solid-phase extraction (SPE) devices are designed for sample preparation. They are manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines, and are suitable for both manual and automated solid-phase extraction.
  • Designed as convenient, reproducible, and disposable solid-phase extraction (SPE) devices
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines
  • Suitable for both manual and automated solid-phase extraction
  • Chemistry: Florisil
  • Barrel size: 3 cc
  • Sorbent weight: 500 mg
  • Particle size: 50-200 μm
  • Format: Vac cartridge
  • Mass Spec compatible
  • Normal-phase mode
  • Pore size: 60 Å
  • Water wettable
  • Units per package: 50
Catalog No. 50-791-159 Supplier Waters Corp Supplier No. 186004611
These convenient, reproducible, and disposable solid-phase extraction (SPE) devices are designed for sample preparation. They are manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines, and are suitable for both manual and automated solid-phase extraction.
  • Designed as convenient, reproducible, and disposable solid-phase extraction (SPE) devices
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP guidelines
  • Suitable for both manual and automated solid-phase extraction
  • Chemistry: Florisil
  • Barrel size: 3 cc
  • Sorbent weight: 500 mg
  • Particle size: 50-200 μm
  • Format: Vac cartridge
  • Mass Spec compatible
  • Normal-phase mode
  • Pore size: 60 Å
  • Water wettable
  • Units per package: 50