Learn More
Waters Corp Waters Vac Cartridge, Florisil, 6 cc, 1 g Sorbent, 50-200 μm
Catalog No. 50791161 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Certified Sep-Pak Florisil 6 cc Vac Cartridge is designed for fieldwork to easily extract solvents, solutions, and samples, facilitating convenient transfer without spillage or sample loss due to evaporation. It is a precision chromatographic tool for sample enrichment and purification.
  • Facilitates easy extraction and convenient transfer of solvents, solutions, and samples in fieldwork.
  • Features Florisil™, an extremely polar, highly active, and weakly basic sorbent for low to moderate polarity species.
  • Suitable for pesticide analysis and adheres to AOAC, EPA, and JPMHLW regulatory methods.
  • Product integrity maintained by high-quality, reproducible packings sealed in a non-contaminating poly-foil pouch.
  • Includes a 20 μm frit for excellent solvent resistance, low extractables, good flow, and depth filtration.
Catalog No. 50-791-161 Supplier Waters Corp Supplier No. 186004613
The Certified Sep-Pak Florisil 6 cc Vac Cartridge is designed for fieldwork to easily extract solvents, solutions, and samples, facilitating convenient transfer without spillage or sample loss due to evaporation. It is a precision chromatographic tool for sample enrichment and purification.
  • Facilitates easy extraction and convenient transfer of solvents, solutions, and samples in fieldwork.
  • Features Florisil™, an extremely polar, highly active, and weakly basic sorbent for low to moderate polarity species.
  • Suitable for pesticide analysis and adheres to AOAC, EPA, and JPMHLW regulatory methods.
  • Product integrity maintained by high-quality, reproducible packings sealed in a non-contaminating poly-foil pouch.
  • Includes a 20 μm frit for excellent solvent resistance, low extractables, good flow, and depth filtration.