Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Certified Sep-Pak Vac Florisil SPE Cartridge, 6 cc, 500 mg

Catalog No. 50791160 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Certified Sep-Pak Vac Florisil SPE (Solid-Phase Extraction) Cartridge is a reproducible, disposable for sample preparation, for high-quality analytical applications.

  • Certified quality: Manufactured in an ISO 13485 and ISO 9002-registered facility, cGMP compliant for FDA Class 1 Medical Devices, ensuring reliability.
  • Florisil chemistry: Normal-phase sorbent.
  • Optimal dimensions: 6 cc, 500 mg sorbent mass.
  • Packaging: Box contains 30 cartridges.
  • Versatile application: Suitable for manual and automated solid-phase extraction.
  • Sample preparation capabilities:
    • Purification: Retains components of interest or interferences.
    • Trace enrichment/concentration: Concentrates analytes from dilute solutions.
    • Fractionation: Isolates analytes based on polarity differences.
    • Solvent exchange: Adsorbs analyte and elutes with compatible solvent.

Catalog No. 50-791-160 Supplier Waters Corp Supplier No. 186004612
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Certified Sep-Pak Vac Florisil SPE (Solid-Phase Extraction) Cartridge is a reproducible, disposable for sample preparation, for high-quality analytical applications.

  • Certified quality: Manufactured in an ISO 13485 and ISO 9002-registered facility, cGMP compliant for FDA Class 1 Medical Devices, ensuring reliability.
  • Florisil chemistry: Normal-phase sorbent.
  • Optimal dimensions: 6 cc, 500 mg sorbent mass.
  • Packaging: Box contains 30 cartridges.
  • Versatile application: Suitable for manual and automated solid-phase extraction.
  • Sample preparation capabilities:
    • Purification: Retains components of interest or interferences.
    • Trace enrichment/concentration: Concentrates analytes from dilute solutions.
    • Fractionation: Isolates analytes based on polarity differences.
    • Solvent exchange: Adsorbs analyte and elutes with compatible solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.