Learn More
Waters Corp Certified Sep-Pak Vac Cartridge, Silica, 6 cc, 500 mg, 55–105 μm
Catalog No. 50791164 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These vac cartridges contain a polar sorbent for adsorbing analytes from non-polar solvents. They also function as a cation exchanger in aqueous mediums and are suitable for vacuum manifolds and automated SPE instruments.
  • Quality controlled for reproducible separations and ultra-low-level detection.
  • Reduces background interferences.
  • Product integrity maintained by poly-foil packaging.
  • Enables quick extraction, preventing spills and sample loss.
  • Precision tool for sample enrichment and purification.
  • High-quality packings ensure reproducible activity and capacity.
  • Superior surface area, pore size, and particle size distributions.
  • 20 μm frit offers favorable flow, low extractables, and strong solvent resistance.
  • Manufactured in a cGMP, ISO 9001-certified facility.
Catalog No. 50-791-164 Supplier Waters Corp Supplier No. 186004616
These vac cartridges contain a polar sorbent for adsorbing analytes from non-polar solvents. They also function as a cation exchanger in aqueous mediums and are suitable for vacuum manifolds and automated SPE instruments.
  • Quality controlled for reproducible separations and ultra-low-level detection.
  • Reduces background interferences.
  • Product integrity maintained by poly-foil packaging.
  • Enables quick extraction, preventing spills and sample loss.
  • Precision tool for sample enrichment and purification.
  • High-quality packings ensure reproducible activity and capacity.
  • Superior surface area, pore size, and particle size distributions.
  • 20 μm frit offers favorable flow, low extractables, and strong solvent resistance.
  • Manufactured in a cGMP, ISO 9001-certified facility.