Learn More
Interscience Certified Standard Weight
SDP
Catalog No. 16948914 Shop All Interscience Products
Change view
Click to view available options
For Use With (Equipment):
DiluFlow
Catalog No. For Use With (Equipment)
16-948-914 DiluFlow
1 options
Catalog No. 16-948-914 Supplier Interscience Supplier No. 513005
Add to Cart
Add to Cart
500 g COFRAC calibrated weight, type F1 for DiluFlow
  • 500 g COFRAC calibrated weight
  • Type F1
  • For use with DiluFlow
Specifications
Product Type Tube Weight
Autoclavable Autoclavable
For Use With (Equipment) DiluFlow
Quantity One 500 g Weight
Sterility Non-sterile