Learn More
Waters Corp Waters Total Recovery Vial, LCGC Certified, Clear Glass, 1 mL, Screw Neck
Catalog No. 50782559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This LCGC Certified Clear Glass Total Recovery Vial is designed for sample preparation and autosampler use. It features a 1 mL volume and a screw neck with a polyethylene septumless cap, providing high cleanliness and chemical inertness for accurate analytical results.
  • Tested for cleanliness by HPLC.
  • Manufactured to tightest dimensional tolerances.
  • Secures sensitive samples and solvents.
  • Chemically inert for interference-free analysis.
  • Compositionally stable to withstand chemical and mechanical forces.
  • No tools required for equipment assembly.
  • Cap manufactured with chemically inert materials.
  • Provides a secure fit to analytical equipment.
  • Offers protection against leakage.
  • Allows optimal visibility of chemicals being analyzed.
Catalog No. 50-782-559 Supplier Waters Corp Supplier No. 186004167C
This LCGC Certified Clear Glass Total Recovery Vial is designed for sample preparation and autosampler use. It features a 1 mL volume and a screw neck with a polyethylene septumless cap, providing high cleanliness and chemical inertness for accurate analytical results.
  • Tested for cleanliness by HPLC.
  • Manufactured to tightest dimensional tolerances.
  • Secures sensitive samples and solvents.
  • Chemically inert for interference-free analysis.
  • Compositionally stable to withstand chemical and mechanical forces.
  • No tools required for equipment assembly.
  • Cap manufactured with chemically inert materials.
  • Provides a secure fit to analytical equipment.
  • Offers protection against leakage.
  • Allows optimal visibility of chemicals being analyzed.