Learn More
Sigma Aldrich Fine Chemicals Biosciences Certified Vial Kit Low Ads
Catalog No. 502900530
Encompass
Supelcos new Low Adsorption (LA) vials are manufactured using a process that decreases the number of hydroxyl groups on the vials glass surface significantly reducing surface activity while improving analytical quantitation and minimizing pH shifts in the sample. This same process also removes unwanted surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis. Unlike other methods used to decrease vial surface activity the elimination of surface activity in LA vials is integral to the manufacturing process and is not a chemical surface treatment.These vials and closures are designed engineered and manufactured for optimum performance in HPLC GC/MS and LC/MS applications. All of the parts of the vial system (vial cap and septum) can come into contact with the sample and are critical to the outcome of the analysis. We recommend using LA vials with MSQ polypropylene cap/PTfE silicone septa. This cap and septa combination was purposely d
Catalog No. 50-290-0530 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 29652U
Supelcos new Low Adsorption (LA) vials are manufactured using a process that decreases the number of hydroxyl groups on the vials glass surface significantly reducing surface activity while improving analytical quantitation and minimizing pH shifts in the sample. This same process also removes unwanted surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis. Unlike other methods used to decrease vial surface activity the elimination of surface activity in LA vials is integral to the manufacturing process and is not a chemical surface treatment.These vials and closures are designed engineered and manufactured for optimum performance in HPLC GC/MS and LC/MS applications. All of the parts of the vial system (vial cap and septum) can come into contact with the sample and are critical to the outcome of the analysis. We recommend using LA vials with MSQ polypropylene cap/PTfE silicone septa. This cap and septa combination was purposely d