Learn More
Certified Vial Kit, Low Adsorption (LA)
Catalog No. p-7204521 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1.5 mL
2 mL
Material:
Polypropylene Cap, Clear Glass, Natural PTFE, Silicone Septa
Polypropylene Cap, Natural PTFE, Silicone Septa
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Material
11-100-9971 1.5 mL Polypropylene Cap, Clear Glass, Natural PTFE, Silicone Septa
11-101-1019 2 mL Polypropylene Cap, Natural PTFE, Silicone Septa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9971 Supplier MilliporeSigma Supelco Supplier No. 29655U
Only null left
Add to Cart
Add to Cart
Manufactured using a process that decreases the number of hydroxyl groups on the vial's glass surface, significantly reducing surface activity while improving analytical quantitation and minimizing pH shifts in the sample.
  • The process removes unwanted surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis.
  • The elimination of surface activity in LA vials is integral to the manufacturing process and is not a chemical surface treatment.
  • These vials and closures are designed, engineered, and manufactured for optimum performance in HPLC, GC/MS, and LC/MS applications.
  • All of the parts of the vial system (vial, cap, and septum) can come into contact with the sample and are critical to the outcome of the analysis.
  • Use LA vials with MSQ polypropylene cap/PTFE silicone septa.
  • This cap and septa combination was purposely designed for use with mass spectrometry.
  • It shows little to no background contamination when compared to other cap and septa products in the marketplace.
TRUSTED_SUSTAINABILITY
Specifications
Color Light Blue
Material Polypropylene Cap, Clear Glass, Natural PTFE, Silicone Septa
Volume (Metric) 1.5 mL
No. per Pack 100
Closure Size 9 mm
Marking Spot No
Product Type CD (Center Draining) Vial
CD is a trademark of QIS, Inc.