Learn More
MSC Excel Dryer Hand Dryer, 8 Second Dry Time, Made in USA Certified, EPD Certified
Catalog No. 502839345
Encompass
The XLERATOR Hand Dryer is an original, patented design, recognized for its high performance and energy efficiency. It offers advanced features such as adjustable speed, sound, and heat controls, providing a hygienic and sustainable hand drying solution. This hand dryer is designed to deliver significant cost savings and reduce environmental impact compared to traditional paper towels.
  • Achieves 95% cost savings compared to paper towels.
  • Includes adjustable speed and sound control.
  • Offers adjustable heat control (high, medium, low, and off).
  • Equipped with an externally visible service LED.
  • Available with electrostatic HEPA filtration for virus removal.
  • Contributes to WELL points and several LEED credits.
  • Reduces carbon footprint by up to 88% compared to paper towels.
  • Proven performance with less than 1% failure rate.
Catalog No. 50-283-9345 Supplier MSC Supplier No. 77671501
The XLERATOR Hand Dryer is an original, patented design, recognized for its high performance and energy efficiency. It offers advanced features such as adjustable speed, sound, and heat controls, providing a hygienic and sustainable hand drying solution. This hand dryer is designed to deliver significant cost savings and reduce environmental impact compared to traditional paper towels.
  • Achieves 95% cost savings compared to paper towels.
  • Includes adjustable speed and sound control.
  • Offers adjustable heat control (high, medium, low, and off).
  • Equipped with an externally visible service LED.
  • Available with electrostatic HEPA filtration for virus removal.
  • Contributes to WELL points and several LEED credits.
  • Reduces carbon footprint by up to 88% compared to paper towels.
  • Proven performance with less than 1% failure rate.