Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precision Dynamics Corp Certis™ B4 Desktop Direct Thermal Label Printer

Catalog No. 21377166
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
21-377-166 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-166 Supplier Precision Dynamics Corp Supplier No. PDB430EA1000
Add to Cart
Add to Cart

High-speed direct thermal printer with a space-saving footprint, purpose-built to meet a variety of healthcare identification

  • Compact, robust design ideal for limited workspaces
  • Prints a wide range of ID solutions: Parent/Infant wristbands, chart labels, medication labels, and more
  • Easy, jam-free media loading for uninterrupted workflow
  • Seamlessly integrates with leading EMRs including Epic, Cerner, Meditech, and others
  • Equipped with USB and Ethernet interfaces for flexible integration with 300 DPI
  • Backed by 3-year warranty coverage

Specifications

Product Type Printer
Color White
Display Type None
For Use With (Equipment) Brady Workstation
Material Plastic
Print Technology Direct Thermal, Thermal Transfer
Length (English) 7.7 in.
Width (English) 6.8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.