Learn More
AdipoGen Cerulenin
SDP
Catalog No. 502562504
Encompass_Preferred
Chemical. CAS 17397-89-6. Formula C12H17NO3. MW 223.3. Isolated from Cephalosporium caerulens. Antibiotic and antifungal compound. Inhibitor of bacterial fatty acid synthesis inhibits FabH, FabB and FabF condensation enzymes. Irreversible fatty acid synthase FAS inhibitor. Inhibits sterol and fatty acid biosynthesis. In fatty acid synthesis, reported to bind in equimolar ratio to beta-keto-acyl-ACP synthase, thus inhibiting protein acylation. In sterol synthesis, inhibits HMG-CoA synthetase activity. Palmitoylation a post-translational lipidation inhibitor. Anticancer agent. Apoptosis inducer in tumor cell lines, with upregulated expression and activity of fatty acid synthase FAS. Shown to induce apoptosis via disruption of the interaction between AIF and Hexokinase II HKII. Necroptosis inhibitor by blocking very long-chain fatty acids VLCFA synthesis and accumulation, which is important in the process of necroptosis RIP1-dependent cell death.
Catalog No. 50-256-2504 Supplier AdipoGen Supplier No. AGCN20513M005
Chemical. CAS 17397-89-6. Formula C12H17NO3. MW 223.3. Isolated from Cephalosporium caerulens. Antibiotic and antifungal compound. Inhibitor of bacterial fatty acid synthesis inhibits FabH, FabB and FabF condensation enzymes. Irreversible fatty acid synthase FAS inhibitor. Inhibits sterol and fatty acid biosynthesis. In fatty acid synthesis, reported to bind in equimolar ratio to beta-keto-acyl-ACP synthase, thus inhibiting protein acylation. In sterol synthesis, inhibits HMG-CoA synthetase activity. Palmitoylation a post-translational lipidation inhibitor. Anticancer agent. Apoptosis inducer in tumor cell lines, with upregulated expression and activity of fatty acid synthase FAS. Shown to induce apoptosis via disruption of the interaction between AIF and Hexokinase II HKII. Necroptosis inhibitor by blocking very long-chain fatty acids VLCFA synthesis and accumulation, which is important in the process of necroptosis RIP1-dependent cell death.