Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerulenin, 98%, Thermo Scientific Chemicals

Catalog No. AAJ64538LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64538LB0 5 mg
AAJ64538MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64538LB0 Supplier Thermo Scientific Chemicals Supplier No. J64538LB0
Add to Cart

Has antibiotic and antifungal properties; inhibits fatty acid and steroid biosynthesis

Cerulenin is used as an antifungal antibiotic that inhibits sterol and fatty acids biosynthesis. Cerulenin induces apoptosis in tumor cell lines. It can be a useful biochemical tool with applications for the study of metabolism and function of fatty acids in the bacterial cell membrane and in yeast. An extensive review including the properties, uses, mechanism of action and structure has been reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerulenin is used as an antifungal antibiotic that inhibits sterol and fatty acids biosynthesis. Cerulenin induces apoptosis in tumor cell lines. It can be a useful biochemical tool with applications for the study of metabolism and function of fatty acids in the bacterial cell membrane and in yeast. An extensive review including the properties, uses, mechanism of action and structure has been reported.

Solubility
Soluble in acetone at 20mg/ml. Also soluble in ethanol, benzene, chloroform, or ethyl acetate.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17397-89-6
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.272
MDL Number MFCD00077686
InChI Key GVEZIHKRYBHEFX-NQQPLRFYSA-N
Synonym (2R,3S,E,E)-2,3-Epoxy-4-oxo-7,10-dodecadienamide
PubChem CID 5282054
ChEBI CHEBI:171741
IUPAC Name (2R,3S)-3-[(4E,7E)-nona-4,7-dienoyl]oxirane-2-carboxamide
SMILES CC=CCC=CCCC(=O)C1C(O1)C(=O)N

Specifications

Type Cerulenin
Physical Form Powder
Color White to Yellow
Quantity 5 mg
Beilstein 4140423
Merck Index 14,2004
Solubility Information Soluble in acetone at 20mg/ml; Also soluble in ethanol,benzene,chloroform,or ethyl acetate.
Formula Weight 223.27
Percent Purity 98%
Chemical Name or Material Cerulenin
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 241-424-8
RTECSNumber JR1670000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.