Learn More
TARGETMOL CHEMICALS INC CERULETIDE 5MG
SDP
Catalog No. 502581268
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Ceruletide (Caerulein) is a potent agonist of cholecystokinin receptor and it also is a safe and effective cholecystokinetic agent. It has a direct spasmogenic effect on the gallbladder muscle and bile ducts. purity: 97%
Catalog No. 50-258-1268 Supplier TARGETMOL CHEMICALS INC Supplier No. T14932
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Ceruletide (Caerulein) is a potent agonist of cholecystokinin receptor and it also is a safe and effective cholecystokinetic agent. It has a direct spasmogenic effect on the gallbladder muscle and bile ducts. purity: 97%