Learn More
Athens Research And Technology Inc Ceruloplasmin (CP), Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. 50489708
Encompass_Preferred
Ceruloplasmin (CP) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 132 kDa. Ceruloplasmin, Human Plasma: 1 MG; Lyophilized from 50 mM potassium phosphate, pH 6.8, 100 mM KCl and 20 mM E-amino caproic acid + 5 mM EDTA. MW 132,000; EC=1.5; Purity: > =95% by SDS-PAGE. Prepared from human plasma in the Athens Research & Tech lab. Common research applications include plasma protein, binding and biochemical studies.
Catalog No. 50-489-708 Supplier Athens Research And Technology Inc Supplier No. 16160305181MG
Ceruloplasmin (CP) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 132 kDa. Ceruloplasmin, Human Plasma: 1 MG; Lyophilized from 50 mM potassium phosphate, pH 6.8, 100 mM KCl and 20 mM E-amino caproic acid + 5 mM EDTA. MW 132,000; EC=1.5; Purity: > =95% by SDS-PAGE. Prepared from human plasma in the Athens Research & Tech lab. Common research applications include plasma protein, binding and biochemical studies.