Learn More
Enzo Life Sciences Ceruloplasmin (human plasma) (1mg). CAS: 9031-37-2
SDP
Supplier:  Enzo Life Sciences ALX200089M001
Encompass_Preferred
Serum copper transport protein. Serum copper transport and iron-oxidizing protein. Expressed in plasma at concentrations of 1-5µM. Contains six copper centers and is known to oxidize amines in a process coupled to the reduction of molecular oxygen. Has ferroxidase activity that is responsible for the oxidation of ferrous iron to its ferric form, which is necessary for efficient iron efflux from the cell . Has previously been considered a target for nitric oxide (NO) and an inhibitor of endothelial nitric oxide synthase (eNOS/NOS III). Catalyzes S-nitrosothiol formation in cell culture media. Is a NO oxidase and nitrite synthase that determines endocrine NO homeostasis. Alternative name: Ferroxidase. Purity: ≥95% (SDS-PAGE). MW: ~134kDa. Formulation: Lyophilized from 50mM potassium phosphate, pH 6.8, containing 100mM potassium chloride, 20mM ε-aminocaproic acid and 5mM EDTA. Source: Isolated from human plasma.
Catalog No. 50-200-7872