Learn More
Discovery Life Sciences Cervical Cancer Tx Naive PBMC
SDP
Catalog No. 5000804044
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04044 Supplier Discovery Life Sciences Supplier No. 121945
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays