Learn More
Lifeline Cell Technology Cervical Epi Media Kit
SDP
Catalog No. NC1944655
Encompass_Preferred
ReproLife CX Medium Complete Kit. Culture medium for the growth of cervical epithelial cells. LM-0055 ReproLife CX Basal Medium and LS-1116 ReproLife CX LifeFactors Kit
Catalog No. NC1944655 Supplier Lifeline Cell Technology Supplier No. LL0072
ReproLife CX Medium Complete Kit. Culture medium for the growth of cervical epithelial cells. LM-0055 ReproLife CX Basal Medium and LS-1116 ReproLife CX LifeFactors Kit